Assay Details
| Gene Symbol: Ppapdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028000.1 | NP_082276.1 | 6 - 7 | 665 | 120 | |
| NR_121642.1 | 6 - 7 | 625 | 120 | |||
| NR_121643.1 | 6 - 7 | 583 | 120 | |||
| GenBank mRNA | AK007542.1 | - | 3 - 4 | 1269 | 120 | |
| AK009455.1 | - | 5 - 6 | 1275 | 120 | ||
| AK144593.1 | - | 6 - 7 | 650 | 120 | ||
| BC023111.1 | - | 6 - 7 | 612 | 120 | ||
| BC039626.1 | - | 5 - 6 | 480 | 120 | ||
| BC062173.1 | - | 6 - 7 | 598 | 120 | ||
| BC089556.1 | - | 6 - 7 | 567 | 120 | ||
| DV661598.1 | - | 3 - 4 | 315 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2175060 | BC089556.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2175060 | BC099489.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2175060 | DV661598.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2175060 | BC023111.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2175060 | AK144593.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281593 | BC089556.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281593 | BC099489.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281593 | CO044265.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281593 | BC023111.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281593 | AK144593.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281593 | BC089556.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281593 | BC099489.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281593 | CO044265.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281593 | BC023111.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281593 | AK144593.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 465875 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 677009 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 627563 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 185592 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143011 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281593 | BC089556.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281593 | BC099489.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281593 | CO044265.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281593 | BC023111.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281593 | AK144593.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281593 | BC089556.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281593 | BC099489.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281593 | CO044265.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281593 | BC023111.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281593 | AK144593.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2175060 | BC089556.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2175060 | BC099489.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2175060 | DV661598.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2175060 | BC023111.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2175060 | AK144593.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143011 | - | - |
| Whole Mouse Genome G4122A | A_51_P281593 | BC089556.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281593 | BC099489.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281593 | CO044265.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281593 | BC023111.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281593 | AK144593.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281593 | BC089556.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281593 | BC099489.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281593 | CO044265.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281593 | BC023111.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281593 | AK144593.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25728722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06807264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06807265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07426247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07426248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06730059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07426239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07360212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25728709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07048326 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10571043 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571037 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571041 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571044 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571038 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571042 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571039 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571040 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428154_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10571036 | - | - |
| Mouse Genome 430 2.0 Array | 1428154_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1457630_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428154_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1457630_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71910_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71910_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800002481.mm.1 | NM_028000.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800002481.mm.1 | NM_028000.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800002481.mm.1 | TC0800000328.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800002482.mm.1 | NM_028000.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800002482.mm.1 | NM_028000.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800002482.mm.1 | TC0800000328.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430B Array | 1428154_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428154_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 329248 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2884610 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2884610 | - | - |

Ppapdc1b
Genomic Map