Assay Details
| Gene Symbol: Tm2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027194.3 | NP_081470.2 | 1 - 2 | 386 | 87 | |
| GenBank mRNA | AK010545.1 | - | 1 - 2 | 318 | 87 | |
| AK045768.1 | - | 1 - 2 | 345 | 87 | ||
| AK080854.1 | - | 1 - 2 | 314 | 87 | ||
| AK083654.1 | - | 1 - 2 | 307 | 87 | ||
| AK085975.1 | - | 1 - 2 | 338 | 87 | ||
| AK086015.1 | - | 1 - 2 | 324 | 87 | ||
| AK086059.1 | - | 1 - 2 | 340 | 87 | ||
| AK088246.1 | - | 1 - 2 | 338 | 87 | ||
| AK149407.1 | - | 1 - 2 | 315 | 87 | ||
| AK160039.1 | - | 1 - 2 | 323 | 87 | ||
| AK166012.1 | - | 1 - 2 | 339 | 87 | ||
| BC026789.1 | - | 1 - 2 | 313 | 4188168 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tm2d2
Genomic Map