Assay Details
Gene Symbol: Htra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081187.3 | NP_001074656.1 | 7 - 8 | 1272 | 71 | |
GenBank mRNA | AK046614.1 | - | 7 - 8 | 1191 | 71 | |
BC132380.1 | - | 7 - 8 | 1232 | 40130657 | 71 | |
BC145842.1 | - | 7 - 8 | 1232 | 40131145 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
