Assay Details
| Gene Symbol: Stk32b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022416.2 | NP_071861.1 | 5 - 6 | 839 | 65 | |
| GenBank mRNA | AJ250840.1 | - | 5 - 6 | 917 | 65 | |
| AK082468.1 | - | 5 - 6 | 840 | 65 | ||
| BC052404.1 | - | 5 - 6 | 749 | 5715729 | 65 | |
| BC056396.1 | - | 5 - 6 | 825 | 6841272 | 65 | |
| BC058412.1 | - | 5 - 6 | 749 | 5715729 | 65 | |
More Information
Set Membership: |
|

Stk32b
Genomic Map