| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300017233.mm.1 |
NM_001032413.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300017233.mm.1 |
NM_001032413.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300017233.mm.1 |
NM_001032414.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300017233.mm.1 |
NM_001032414.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300017233.mm.1 |
NM_001289600.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300017233.mm.1 |
NM_001289601.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300017233.mm.1 |
NM_028460.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300017233.mm.1 |
NM_028460.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300017233.mm.1 |
NM_152799.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300017233.mm.1 |
NM_152799.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300017233.mm.1 |
TC0300003234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300017234.mm.1 |
NM_001032413.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300017234.mm.1 |
NM_001032413.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300017234.mm.1 |
NM_001032414.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300017234.mm.1 |
NM_001032414.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300017234.mm.1 |
NM_001289600.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300017234.mm.1 |
NM_028460.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300017234.mm.1 |
NM_028460.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300017234.mm.1 |
NM_152799.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300017234.mm.1 |
NM_152799.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300017234.mm.1 |
TC0300003234.mm.1 |
MTA TC |