Assay Details
| Gene Symbol: Add1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024458.3 | NP_001019629.2 | 11 - 12 | 1739 | 68 | |
| NM_001102444.1 | NP_001095914.1 | 11 - 12 | 1832 | 68 | ||
| NM_013457.3 | NP_038485.1 | 11 - 12 | 1739 | 68 | ||
| GenBank mRNA | AF096839.1 | - | 11 - 12 | 1637 | 68 | |
| AF189771.1 | - | 10 - 11 | 1519 | 68 | ||
| AK054558.1 | - | 11 - 12 | 1684 | 68 | ||
| AK179785.1 | - | 2 - 3 | 225 | 68 | ||
| AK189311.1 | - | 2 - 3 | 166 | 68 | ||
| AK190231.1 | - | 1 - 2 | 77 | 68 | ||
| BC034368.1 | - | 10 - 11 | 1570 | 4008924 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Add1
Genomic Map