Assay Details
| Gene Symbol: Add1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024458.3 | NP_001019629.2 | 12 - 13 | 1824 | 84 | |
| NM_001102444.1 | NP_001095914.1 | 12 - 13 | 1917 | 84 | ||
| NM_013457.3 | NP_038485.1 | 12 - 13 | 1824 | 84 | ||
| GenBank mRNA | AF096839.1 | - | 12 - 13 | 1722 | 84 | |
| AF189771.1 | - | 11 - 12 | 1604 | 84 | ||
| AK054558.1 | - | 12 - 13 | 1769 | 84 | ||
| AK189311.1 | - | 3 - 4 | 251 | 84 | ||
| AK217867.1 | - | 3 - 4 | 250 | 84 | ||
| BC034368.1 | - | 11 - 12 | 1655 | 4008924 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Add1
Genomic Map