Assay Details
| Gene Symbol: Add1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024458.3 | NP_001019629.2 | 7 - 8 | 1105 | 110 | |
| NM_001102444.1 | NP_001095914.1 | 7 - 8 | 1105 | 110 | ||
| NM_013457.3 | NP_038485.1 | 7 - 8 | 1105 | 110 | ||
| GenBank mRNA | AF096839.1 | - | 7 - 8 | 1003 | 110 | |
| AF189771.1 | - | 6 - 7 | 885 | 110 | ||
| AK054558.1 | - | 7 - 8 | 957 | 110 | ||
| BC034368.1 | - | 6 - 7 | 936 | 4008924 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Add1
Genomic Map