Assay Details
| Gene Symbol: Clnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013748.3 | NP_038776.3 | 17 - 18 | 1113 | 69 | |
| GenBank mRNA | AB021220.1 | - | 17 - 18 | 1233 | 69 | |
| AF187819.1 | - | 17 - 18 | 1113 | 69 | ||
| BC125637.1 | - | 17 - 18 | 1110 | 40130152 | 69 | |
| BC125639.1 | - | 17 - 18 | 1110 | 40130154 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clnk
Genomic Map