Assay Details
| Gene Symbol: Clnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013748.3 | NP_038776.3 | 7 - 8 | 538 | 71 | |
| GenBank mRNA | AB021220.1 | - | 7 - 8 | 71 | ||
| AF187819.1 | - | 7 - 8 | 538 | 71 | ||
| AK082826.1 | - | 3 - 4 | 276 | 71 | ||
| BC125637.1 | - | 7 - 8 | 40130152 | 71 | ||
| BC125639.1 | - | 7 - 8 | 535 | 40130154 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clnk
Genomic Map