Assay Details
| Gene Symbol: Slc2a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012363.2 | NP_001012363.2 | 7 - 8 | 920 | 81 | |
| NM_001102414.1 | NP_001095884.1 | 7 - 8 | 1119 | 81 | ||
| GenBank mRNA | AF469480.1 | - | 7 - 8 | 1123 | 81 | |
| AK156740.1 | - | 7 - 8 | 1023 | 81 | ||
| AK172234.1 | - | 7 - 8 | 1023 | 81 | ||
| AY776155.1 | - | 7 - 8 | 899 | 81 | ||
| BC138213.1 | - | 8 - 9 | 1055 | 8861232 | 81 | |
| BC145198.1 | - | 9 - 10 | 1055 | 9053749 | 81 | |
More Information
Set Membership: |
|

Slc2a9
Genomic Map