Assay Details
| Gene Symbol: Adgra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133911.1 | NP_598672.1 | 13 - 14 | 2211 | 66 | |
| GenBank mRNA | AK028929.1 | - | 13 - 14 | 2150 | 66 | |
| AK131924.1 | - | 2 - 3 | 353 | 66 | ||
| BC019649.1 | - | 2 - 3 | 338 | 66 | ||
| BC020317.2 | - | 8 - 9 | 1344 | 66 | ||
| BC052391.1 | - | 13 - 14 | 2211 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adgra2
Genomic Map