Assay Details
| Gene Symbol: Adgra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133911.1 | NP_598672.1 | 1 - 2 | 444 | 80 | |
| GenBank mRNA | AK014455.1 | - | 1 - 2 | 416 | 80 | |
| AK028929.1 | - | 1 - 2 | 383 | 80 | ||
| BC052391.1 | - | 1 - 2 | 444 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adgra2
Genomic Map