Assay Details
| Gene Symbol: Il6ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010559.2 | NP_034689.2 | 3 - 4 | 574 | 63 | |
| GenBank mRNA | AK046337.1 | - | 3 - 4 | 573 | 63 | |
| AK082270.1 | - | 3 - 4 | 574 | 63 | ||
| AK141197.1 | - | 3 - 4 | 609 | 63 | ||
| X51975.1 | - | 3 - 4 | 465 | 63 | ||
| X51976.1 | - | 454 | 63 | |||
| X53802.1 | - | 3 - 4 | 574 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il6ra
Genomic Map