Assay Details
| Gene Symbol: Il6ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010559.2 | NP_034689.2 | 5 - 6 | 922 | 69 | |
| GenBank mRNA | AK082270.1 | - | 5 - 6 | 923 | 69 | |
| AK141197.1 | - | 5 - 6 | 957 | 69 | ||
| BC019388.1 | - | 5 - 6 | 916 | 69 | ||
| X51975.1 | - | 5 - 6 | 813 | 69 | ||
| X51976.1 | - | 802 | 69 | |||
| X53802.1 | - | 5 - 6 | 922 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il6ra
Genomic Map