Assay Details
| Gene Symbol: 4933434E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287087.1 | NP_001274016.1 | 2 - 3 | 238 | 101 | |
| GenBank mRNA | AK077215.1 | - | 2 - 3 | 239 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK077215.1, NM_001287087.1 transcripts.
Set Membership: |
|

4933434E20Rik
Genomic Map