Assay Details
| Gene Symbol: 4933434E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287086.1 | NP_001274015.1 | 1 - 2 | 313 | 82 | |
| NM_001287087.1 | NP_001274016.1 | 3 - 4 | 458 | 82 | ||
| NM_025762.3 | NP_080038.1 | 1 - 2 | 313 | 82 | ||
| NM_027500.4 | NP_081776.2 | 1 - 2 | 313 | 82 | ||
| GenBank mRNA | AK017833.1 | - | 1 - 2 | 281 | 82 | |
| AK034749.1 | - | 1 - 2 | 276 | 82 | ||
| AK051238.1 | - | 1 - 2 | 206 | 82 | ||
| AK077215.1 | - | 3 - 4 | 459 | 82 | ||
| AK150324.1 | - | 1 - 2 | 264 | 82 | ||
| AK150423.1 | - | 1 - 2 | 264 | 82 | ||
| AK150831.1 | - | 1 - 2 | 264 | 82 | ||
| AK151002.1 | - | 1 - 2 | 263 | 82 | ||
| AK151263.1 | - | 1 - 2 | 262 | 82 | ||
| AK214769.1 | - | 1 - 2 | 63 | 82 | ||
| AY533135.1 | - | 1 - 2 | 61 | 82 | ||
| BB850680.1 | - | 1 - 2 | 287 | 82 | ||
| BC018438.1 | - | 1 - 2 | 243 | 2609299 | 82 | |
| BC027190.1 | - | 1 - 2 | 173 | 3257131 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC018438.1,NM_001287086.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933434E20Rik
Genomic Map