Assay Details
| Gene Symbol: Ppat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172146.2 | NP_742158.1 | 7 - 8 | 1379 | 103 | |
| GenBank mRNA | AK147750.1 | - | 7 - 8 | 1029 | 103 | |
| AK148034.1 | - | 7 - 8 | 983 | 103 | ||
| AK159141.1 | - | 8 - 9 | 1529 | 103 | ||
| AK159859.1 | - | 8 - 9 | 1529 | 103 | ||
| AK160542.1 | - | 7 - 8 | 1004 | 103 | ||
| AK164506.1 | - | 7 - 8 | 993 | 103 | ||
| AK167057.1 | - | 7 - 8 | 1018 | 103 | ||
| BC023841.1 | - | 7 - 8 | 1006 | 5346105 | 103 | |
More Information
Set Membership: |
|

Ppat
Genomic Map