Assay Details
| Gene Symbol: Gtf3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027901.2 | NP_082177.2 | 7 - 8 | 1394 | 116 | |
| GenBank mRNA | AK004895.1 | - | 7 - 8 | 1172 | 116 | |
| AK035792.1 | - | 7 - 8 | 1160 | 116 | ||
| AK043944.1 | - | 7 - 8 | 1161 | 116 | ||
| AK083585.1 | - | 7 - 8 | 1396 | 116 | ||
| AK083636.1 | - | 6 - 7 | 1149 | 116 | ||
| AK129033.1 | - | 6 - 7 | 1144 | 116 | ||
| AK138760.1 | - | 1163 | 116 | |||
| AK149771.1 | - | 8 - 9 | 1291 | 116 | ||
| AK151300.1 | - | 7 - 8 | 1159 | 116 | ||
| AK152439.1 | - | 7 - 8 | 1159 | 116 | ||
| AK159938.1 | - | 7 - 8 | 1161 | 116 | ||
| BC034369.1 | - | 7 - 8 | 1172 | 3994677 | 116 | |
| BC043100.1 | - | 7 - 8 | 1382 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gtf3c2
Genomic Map