Assay Details
| Gene Symbol: Gtf3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027901.2 | NP_082177.2 | 8 - 9 | 1626 | 69 | |
| GenBank mRNA | AK004895.1 | - | 8 - 9 | 1404 | 69 | |
| AK012095.1 | - | 1 - 2 | 223 | 69 | ||
| AK043944.1 | - | 8 - 9 | 1393 | 69 | ||
| AK083585.1 | - | 8 - 9 | 1628 | 69 | ||
| AK083636.1 | - | 7 - 8 | 1381 | 69 | ||
| AK129033.1 | - | 7 - 8 | 1376 | 69 | ||
| AK138760.1 | - | 1395 | 69 | |||
| AK149771.1 | - | 9 - 10 | 1523 | 69 | ||
| AK151300.1 | - | 8 - 9 | 1391 | 69 | ||
| AK152439.1 | - | 8 - 9 | 1391 | 69 | ||
| AK159938.1 | - | 8 - 9 | 1394 | 69 | ||
| BC034369.1 | - | 8 - 9 | 1404 | 3994677 | 69 | |
| BC043100.1 | - | 8 - 9 | 1614 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gtf3c2
Genomic Map