Assay Details
| Gene Symbol: Ccl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011331.2 | NP_035461.2 | 2 - 3 | 253 | 75 | |
| GenBank mRNA | AF065934.1 | - | 2 - 3 | 219 | 75 | |
| AF065935.1 | - | 2 - 3 | 219 | 75 | ||
| AF065936.1 | - | 2 - 3 | 219 | 75 | ||
| AF065937.1 | - | 2 - 3 | 219 | 75 | ||
| AF065938.1 | - | 2 - 3 | 219 | 75 | ||
| AK012356.1 | - | 2 - 3 | 253 | 75 | ||
| BC027520.1 | - | 2 - 3 | 208 | 1548072 | 75 | |
| U50712.1 | - | 2 - 3 | 252 | 75 | ||
| U66670.1 | - | 2 - 3 | 229 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
angiogenesis
monocyte chemotaxis
positive regulation of leukocyte migration
chemotaxis
inflammatory response
immune response
cytoskeleton organization
regulation of cell shape
cytokine-mediated signaling pathway
lipopolysaccharide-mediated signaling pathway
negative regulation of glial cell apoptotic process
protein kinase B signaling
negative regulation of neuron apoptotic process
astrocyte cell migration
macrophage chemotaxis
negative regulation of leukocyte proliferation
cellular response to organic cyclic compound
positive regulation of calcium ion import
negative regulation of natural killer cell chemotaxis
cytokine activity
chemokine activity
CCR2 chemokine receptor binding
MAPK cascade
angiogenesis
monocyte chemotaxis
positive regulation of leukocyte migration
chemotaxis
inflammatory response
immune response
cytoskeleton organization
regulation of cell shape
cytokine-mediated signaling pathway
lipopolysaccharide-mediated signaling pathway
negative regulation of glial cell apoptotic process
protein kinase B signaling
negative regulation of neuron apoptotic process
astrocyte cell migration
macrophage chemotaxis
negative regulation of leukocyte proliferation
cellular response to organic cyclic compound
positive regulation of calcium ion import
negative regulation of natural killer cell chemotaxis
cytokine activity
chemokine activity
CCR2 chemokine receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984556 | NM_011331.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984556 | AK012356.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984556 | BC027520.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984556 | U66670.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604309 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174813 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167233 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179971 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984556 | NM_011331.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984556 | AK012356.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984556 | BC027520.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984556 | U66670.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P249514 | NM_011331.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P249514 | AF065934.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P249514 | AF065935.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P249514 | AF065936.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P249514 | AF065937.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27964319 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174813 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167233 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179971 | - | - |
| Whole Mouse Genome G4122A | A_52_P249514 | NM_011331.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P249514 | AF065934.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P249514 | AF065935.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P249514 | AF065936.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P249514 | AF065937.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127909 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167233 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174813 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179971 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09253434 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09253421 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09253438 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09253425 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09253419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27964331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09253411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09253425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09715886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09253439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09253408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09253404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09253436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09679474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09253413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27964314 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10379531 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379533 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379534 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379532 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419282_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10379530 | - | - |
| Mouse Genome 430 2.0 Array | 1419282_at | - | - |
| Mouse Genome 430 2.0 Array | 1419282_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20293_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20293_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100011467.mm.1 | NM_011331.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100011467.mm.1 | NM_011331.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100011467.mm.1 | TC1100001222.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100011468.mm.1 | NM_011331.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100011468.mm.1 | NM_011331.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100011468.mm.1 | TC1100001222.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419282_PM_at | - | - |
| Mouse Expression Set 430 | 1419282_at | - | - |
| Murine Genome U74v2 Set | 93717_PM_at | - | - |
| Murine Genome U74v2 Set | 93717_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419282_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419282_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 913289 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 7000592 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 7000592 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007000592 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2771766 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161805 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3161805 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2771766 | - | - |
| mouse-6 v1.1 (Illumina) | scl41157.3.1_54-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_6755419-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7000592 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41157.3.1_54-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7000592 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2771766 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3161805 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2771766 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3161805 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | GI_6755419-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl41157.3.1_54-S | - | - |

Ccl12
Genomic Map