Assay Details
| Gene Symbol: Fam178a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081225.1 | NP_001074694.1 | 5 - 6 | 2133 | 82 | |
| GenBank mRNA | AK030951.1 | - | 1 - 2 | 449 | 82 | |
| AK031688.1 | - | 5 - 6 | 2105 | 82 | ||
| AK036700.1 | - | 3 - 4 | 1710 | 82 | ||
| BC150999.1 | - | 5 - 6 | 1986 | 9087912 | 82 | |
| BC157937.1 | - | 5 - 6 | 1986 | 9088117 | 82 | |
| BC172097.1 | - | 5 - 6 | 1986 | 9054383 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK030951.1 transcripts.
Set Membership: |
|

Fam178a
Genomic Map