Assay Details
| Gene Symbol: Fam178a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081225.1 | NP_001074694.1 | 6 - 7 | 2206 | 79 | |
| GenBank mRNA | AK028324.1 | - | 2 - 3 | 156 | 79 | |
| AK030951.1 | - | 2 - 3 | 522 | 79 | ||
| AK031688.1 | - | 6 - 7 | 2178 | 79 | ||
| AK036700.1 | - | 4 - 5 | 1783 | 79 | ||
| AK147588.1 | - | 6 - 7 | 2679 | 79 | ||
| AK161809.1 | - | 2 - 3 | 569 | 79 | ||
| BC060679.1 | - | 2 - 3 | 123 | 79 | ||
| BC150999.1 | - | 6 - 7 | 2059 | 9087912 | 79 | |
| BC157937.1 | - | 6 - 7 | 2059 | 9088117 | 79 | |
| BC172097.1 | - | 6 - 7 | 2059 | 9054383 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK030951.1 transcripts.
Set Membership: |
|

Fam178a
Genomic Map