Assay Details
| Gene Symbol: Nt5dc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027289.1 | NP_081565.1 | 7 - 8 | 584 | 75 | |
| GenBank mRNA | BC011230.1 | - | 7 - 8 | 584 | 4207917 | 75 |
More Information
Updated Mapping Information
Set Membership: |
|

Nt5dc2
Genomic Map