Assay Details
| Gene Symbol: Nt5dc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027289.1 | NP_081565.1 | 11 - 12 | 998 | 63 | |
| GenBank mRNA | AK010966.1 | - | 2 - 3 | 260 | 63 | |
| BC011230.1 | - | 11 - 12 | 998 | 4207917 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509871 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509886 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509869 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509885 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509884 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509868 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509889 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509890 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509872 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509887 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509883 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509958 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509960 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509959 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296796 | NM_027289.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296796 | BC011230.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296796 | NM_027289.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296796 | BC011230.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482791 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 236724 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 839278 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147207 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152113 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296796 | NM_027289.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296796 | BC011230.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296796 | NM_027289.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296796 | BC011230.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152113 | - | - |
| Whole Mouse Genome G4122A | A_51_P296796 | NM_027289.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P296796 | BC011230.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P296796 | NM_027289.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P296796 | BC011230.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152113 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11283241 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11283239 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11283256 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11283246 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11283180 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11283251 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509872 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509883 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509887 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509869 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509889 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509958 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509960 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509885 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509871 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509884 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509890 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509886 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509868 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29509959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29509913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11802703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509880 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11591386 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11802706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11273890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509907 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509893 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509845 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11283244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11189859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11243857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11775911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11283227 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11283254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11283222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509865 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11283239 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11273877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11802702 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11727875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11283249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11283200 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11283233 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11283241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509868 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11283256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29509961 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10413723 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413722 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413715 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413719 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413724 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413716 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413721 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413718 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413714 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413713 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413717 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413720 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413725 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424882_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10413710 | - | - |
| Mouse Genome 430 2.0 Array | 1424882_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1424882_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70021_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70021_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400002766.mm.1 | NM_027289.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002766.mm.1 | NM_027289.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002766.mm.1 | TC1400000322.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424882_a_at | - | - |
| Mouse Expression Set 430 | 1424882_PM_a_at | - | - |
| Murine Genome U74v2 Set | 96885_PM_at | - | - |
| Murine Genome U74v2 Set | 96885_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424882_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424882_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424882_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424882_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 105080047 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105080047 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2660519 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2660519 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105080047 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105080047 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2544343 | - | - |

Nt5dc2
Genomic Map