Assay Details
| Gene Symbol: Atp10d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153389.3 | NP_700438.3 | 13 - 14 | 2794 | 65 | |
| NR_003966.1 | 13 - 14 | 2808 | 65 | |||
| GenBank mRNA | AJ441079.1 | - | 13 - 14 | 2794 | 65 | |
| BC141328.1 | - | 13 - 14 | 2645 | 9055940 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp10d
Genomic Map