Assay Details
| Gene Symbol: Insr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010568.2 | NP_034698.2 | 12 - 13 | 3143 | 101 | |
| GenBank mRNA | J05149.1 | - | 12 - 13 | 2702 | 101 | |
| KC356871.1 | - | 13 - 14 | 2690 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Insr
Genomic Map