Assay Details
| Gene Symbol: Insr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010568.2 | NP_034698.2 | 3 - 4 | 1458 | 63 | |
| GenBank mRNA | J05149.1 | - | 3 - 4 | 1017 | 63 | |
| KC356871.1 | - | 3 - 4 | 969 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Insr
Genomic Map