Assay Details
| Gene Symbol: Insr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010568.2 | NP_034698.2 | 8 - 9 | 2355 | 155 | |
| GenBank mRNA | EU346716.1 | - | 1 - 2 | 171 | 155 | |
| J05149.1 | - | 8 - 9 | 1914 | 155 | ||
| KC356871.1 | - | 8 - 9 | 1866 | 155 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Insr
Genomic Map