Assay Details
| Gene Symbol: Kit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122733.1 | NP_001116205.1 | 13 - 14 | 2067 | 99 | |
| NM_021099.3 | NP_066922.2 | 13 - 14 | 2051 | 99 | ||
| GenBank mRNA | AK046795.1 | - | 13 - 14 | 2059 | 99 | |
| AK047010.1 | - | 13 - 14 | 2064 | 99 | ||
| AK145462.1 | - | 10 - 11 | 1292 | 99 | ||
| AY536430.1 | - | 13 - 14 | 2014 | 99 | ||
| AY536431.1 | - | 13 - 14 | 2014 | 99 | ||
| BC026713.1 | - | 13 - 14 | 2046 | 99 | ||
| BC052457.1 | - | 13 - 14 | 2026 | 6830637 | 99 | |
| BC075716.1 | - | 13 - 14 | 2052 | 3673641 | 99 | |
| Y00864.1 | - | 13 - 14 | 2018 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kit
Genomic Map