Assay Details
| Gene Symbol: Kit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122733.1 | NP_001116205.1 | 18 - 19 | 2667 | 113 | |
| NM_021099.3 | NP_066922.2 | 18 - 19 | 2651 | 113 | ||
| GenBank mRNA | AK046795.1 | - | 18 - 19 | 2659 | 113 | |
| AK047010.1 | - | 18 - 19 | 2664 | 113 | ||
| AK145462.1 | - | 15 - 16 | 1892 | 113 | ||
| AY536430.1 | - | 18 - 19 | 2614 | 113 | ||
| BC026713.1 | - | 18 - 19 | 2646 | 113 | ||
| BC052457.1 | - | 18 - 19 | 2626 | 6830637 | 113 | |
| BC075716.1 | - | 18 - 19 | 2652 | 3673641 | 113 | |
| EF472963.1 | - | 2 - 3 | 271 | 113 | ||
| X65997.1 | - | 2 - 3 | 686 | 113 | ||
| Y00864.1 | - | 18 - 19 | 2618 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kit
Genomic Map