Assay Details
| Gene Symbol: Kit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122733.1 | NP_001116205.1 | 3 - 4 | 682 | 67 | |
| NM_021099.3 | NP_066922.2 | 3 - 4 | 678 | 67 | ||
| GenBank mRNA | AK046795.1 | - | 3 - 4 | 674 | 67 | |
| AK047010.1 | - | 3 - 4 | 678 | 67 | ||
| AY536430.1 | - | 3 - 4 | 641 | 67 | ||
| AY536431.1 | - | 3 - 4 | 641 | 67 | ||
| BC026713.1 | - | 3 - 4 | 661 | 67 | ||
| BC052457.1 | - | 3 - 4 | 653 | 6830637 | 67 | |
| BC075716.1 | - | 3 - 4 | 667 | 3673641 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kit
Genomic Map