Assay Details
| Gene Symbol: Kit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122733.1 | NP_001116205.1 | 6 - 7 | 1185 | 77 | |
| NM_021099.3 | NP_066922.2 | 6 - 7 | 1181 | 77 | ||
| GenBank mRNA | AK046795.1 | - | 6 - 7 | 1177 | 77 | |
| AK047010.1 | - | 6 - 7 | 1181 | 77 | ||
| AK145462.1 | - | 3 - 4 | 422 | 77 | ||
| AY536430.1 | - | 6 - 7 | 1144 | 77 | ||
| AY536431.1 | - | 6 - 7 | 1144 | 77 | ||
| BC026713.1 | - | 6 - 7 | 1164 | 77 | ||
| BC052457.1 | - | 6 - 7 | 1156 | 6830637 | 77 | |
| BC075716.1 | - | 6 - 7 | 1170 | 3673641 | 77 | |
| Y00864.1 | - | 6 - 7 | 1148 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kit
Genomic Map