Assay Details
| Gene Symbol: Kat6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081149.1 | NP_001074618.1 | 13 - 14 | 2486 | 63 | |
| GenBank mRNA | AK028058.1 | - | 12 - 13 | 2405 | 63 | |
| AK036885.1 | - | 13 - 14 | 2481 | 63 | ||
| AK082463.1 | - | 13 - 14 | 2489 | 63 | ||
| AK132474.1 | - | 12 - 13 | 2394 | 63 | ||
| AK143408.1 | - | 13 - 14 | 2477 | 63 | ||
| BC024786.1 | - | 5 - 6 | 612 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kat6a
Genomic Map