Assay Details
| Gene Symbol: Kat6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081149.1 | NP_001074618.1 | 4 - 5 | 1203 | 98 | |
| GenBank mRNA | AK028058.1 | - | 3 - 4 | 1122 | 98 | |
| AK036885.1 | - | 4 - 5 | 1198 | 98 | ||
| AK039615.1 | - | 4 - 5 | 1201 | 98 | ||
| AK054322.1 | - | 4 - 5 | 1197 | 98 | ||
| AK082463.1 | - | 4 - 5 | 1206 | 98 | ||
| AK132474.1 | - | 3 - 4 | 1111 | 98 | ||
| AK143408.1 | - | 4 - 5 | 1194 | 98 | ||
More Information
Set Membership: |
|

Kat6a
Genomic Map