Assay Details
| Gene Symbol: Kat6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081149.1 | NP_001074618.1 | 6 - 7 | 1398 | 85 | |
| GenBank mRNA | AK028058.1 | - | 5 - 6 | 1317 | 85 | |
| AK036885.1 | - | 6 - 7 | 1393 | 85 | ||
| AK039615.1 | - | 6 - 7 | 1396 | 85 | ||
| AK054322.1 | - | 6 - 7 | 1392 | 85 | ||
| AK082463.1 | - | 6 - 7 | 1401 | 85 | ||
| AK132474.1 | - | 5 - 6 | 1306 | 85 | ||
| AK143408.1 | - | 6 - 7 | 1389 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kat6a
Genomic Map