Assay Details
| Gene Symbol: Plat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008872.2 | NP_032898.2 | 1 - 2 | 82 | 63 | |
| GenBank mRNA | AK053476.1 | - | 1 - 2 | 99 | 63 | |
| AK135857.1 | - | 1 - 2 | 73 | 63 | ||
| AK135965.1 | - | 1 - 2 | 74 | 63 | ||
| AK141665.1 | - | 1 - 2 | 73 | 63 | ||
| AK159620.1 | - | 1 - 2 | 73 | 63 | ||
| AK166639.1 | - | 1 - 2 | 73 | 63 | ||
| BC011256.1 | - | 1 - 2 | 79 | 4038404 | 63 | |
| BC057967.1 | - | 1 - 2 | 62 | 4979664 | 63 | |
| BC061508.1 | - | 1 - 2 | 63 | 5352018 | 63 | |
| CJ151826.1 | - | 1 - 2 | 95 | 63 | ||
| J03520.1 | - | 1 - 2 | 65 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK135857.1, AK135965.1, BC011256.1, BC057967.1, BC061508.1, J03520.1, NM_008872.2 transcripts.
Set Membership: |
|

Plat
Genomic Map