Assay Details
| Gene Symbol: Slain2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113423.2 | NP_001106894.1 | 2 - 3 | 797 | 141 | |
| NM_153567.3 | NP_705795.2 | 2 - 3 | 797 | 141 | ||
| GenBank mRNA | AK039970.1 | - | 2 - 3 | 742 | 141 | |
| AK129365.1 | - | 2 - 3 | 786 | 141 | ||
| AK146432.1 | - | 2 - 3 | 796 | 141 | ||
| BC010836.1 | - | 2 - 3 | 785 | 141 | ||
| BC076573.1 | - | 2 - 3 | 730 | 141 | ||
| CF728452.1 | - | 3 - 4 | 290 | 141 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slain2
Genomic Map