Assay Details
| Gene Symbol: Slain2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113423.2 | NP_001106894.1 | 3 - 4 | 969 | 78 | |
| NM_153567.3 | NP_705795.2 | 3 - 4 | 969 | 78 | ||
| GenBank mRNA | AK039970.1 | - | 3 - 4 | 914 | 78 | |
| AK129365.1 | - | 3 - 4 | 958 | 78 | ||
| AK146432.1 | - | 3 - 4 | 968 | 78 | ||
| BC010836.1 | - | 3 - 4 | 957 | 78 | ||
| BC019454.1 | - | 5 - 6 | 78 | |||
| BC038019.1 | - | 1 - 2 | 174 | 3597423 | 78 | |
| BC076573.1 | - | 3 - 4 | 902 | 78 | ||
| CF728452.1 | - | 4 - 5 | 462 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC038019.1 transcripts.
Set Membership: |
|

Slain2
Genomic Map