Assay Details
| Gene Symbol: Lnx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080795.4 | NP_542985.4 | 5 - 6 | 1433 | 85 | |
| GenBank mRNA | AF401681.1 | - | 5 - 6 | 1390 | 85 | |
| AK036218.1 | - | 5 - 6 | 1434 | 85 | ||
| AK047053.1 | - | 5 - 6 | 1417 | 85 | ||
| AK140320.1 | - | 5 - 6 | 1368 | 85 | ||
| AK170146.1 | - | 5 - 6 | 1394 | 85 | ||
| BC117937.1 | - | 5 - 6 | 1388 | 40097802 | 85 | |
| BC117938.1 | - | 5 - 6 | 1387 | 40097810 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lnx2
Genomic Map