Assay Details
| Gene Symbol: Xab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026156.2 | NP_080432.1 | 14 - 15 | 1988 | 56 | |
| GenBank mRNA | AK002890.1 | - | 14 - 15 | 1989 | 56 | |
| AK031533.1 | - | 2075 | 56 | |||
| AK087952.1 | - | 11 - 12 | 2251 | 56 | ||
| AK132434.1 | - | 14 - 15 | 2002 | 56 | ||
| AK145925.1 | - | 14 - 15 | 1990 | 56 | ||
| BC021341.1 | - | 14 - 15 | 1986 | 5066146 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xab2
Genomic Map