Assay Details
| Gene Symbol: Xab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026156.2 | NP_080432.1 | 7 - 8 | 985 | 64 | |
| GenBank mRNA | AK002890.1 | - | 7 - 8 | 986 | 64 | |
| AK031533.1 | - | 995 | 64 | |||
| AK087952.1 | - | 7 - 8 | 992 | 64 | ||
| AK132434.1 | - | 7 - 8 | 999 | 64 | ||
| AK145925.1 | - | 7 - 8 | 987 | 64 | ||
| BC021341.1 | - | 7 - 8 | 983 | 5066146 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xab2
Genomic Map