Assay Details
| Gene Symbol: Timm44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011592.2 | NP_035722.2 | 10 - 11 | 1056 | 105 | |
| GenBank mRNA | AK153866.1 | - | 10 - 11 | 1061 | 105 | |
| BC023454.1 | - | 10 - 11 | 1032 | 105 | ||
| BC110677.1 | - | 10 - 11 | 1054 | 4979986 | 105 | |
| BC117523.1 | - | 10 - 11 | 1046 | 40105563 | 105 | |
| BC117524.1 | - | 10 - 11 | 1046 | 40105561 | 105 | |
| U69898.1 | - | 10 - 11 | 1046 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Timm44
Genomic Map