Assay Details
Gene Symbol: Timm44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011592.2 | NP_035722.2 | 1 - 2 | 64 | 100 | |
GenBank mRNA | CA493459.1 | - | 1 - 2 | 102 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_011592.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
