Assay Details
| Gene Symbol: Timm44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011592.2 | NP_035722.2 | 5 - 6 | 563 | 63 | |
| GenBank mRNA | AK153866.1 | - | 5 - 6 | 567 | 63 | |
| BC023454.1 | - | 5 - 6 | 539 | 63 | ||
| BC043479.1 | - | 5 - 6 | 543 | 63 | ||
| BC110677.1 | - | 5 - 6 | 561 | 4979986 | 63 | |
| BC117523.1 | - | 5 - 6 | 553 | 40105563 | 63 | |
| BC117524.1 | - | 5 - 6 | 553 | 40105561 | 63 | |
| BC128353.1 | - | 1 - 2 | 63 | 63 | ||
| BC139766.1 | - | 1 - 2 | 63 | 63 | ||
| U69898.1 | - | 5 - 6 | 553 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Timm44
Genomic Map