Assay Details
| Gene Symbol: Stxbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011503.4 | NP_035633.1 | 3 - 4 | 184 | 92 | |
| GenBank mRNA | AF263345.1 | - | 3 - 4 | 191 | 92 | |
| AK088364.1 | - | 3 - 4 | 206 | 92 | ||
| AK133419.1 | - | 3 - 4 | 185 | 92 | ||
| BC003477.1 | - | 3 - 4 | 174 | 92 | ||
| BC008102.1 | - | 3 - 4 | 194 | 3966319 | 92 | |
| D42068.1 | - | 3 - 4 | 176 | 92 | ||
| U19520.1 | - | 3 - 4 | 198 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stxbp2
Genomic Map