Assay Details
| Gene Symbol: Stxbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011503.4 | NP_035633.1 | 7 - 8 | 586 | 83 | |
| GenBank mRNA | AF263345.1 | - | 7 - 8 | 593 | 83 | |
| AK088364.1 | - | 7 - 8 | 608 | 83 | ||
| AK133419.1 | - | 7 - 8 | 587 | 83 | ||
| BC003477.1 | - | 7 - 8 | 576 | 83 | ||
| BC008102.1 | - | 7 - 8 | 596 | 3966319 | 83 | |
| D42068.1 | - | 7 - 8 | 578 | 83 | ||
| U19520.1 | - | 7 - 8 | 600 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stxbp2
Genomic Map