Assay Details
| Gene Symbol: Cers4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026058.4 | NP_080334.3 | 2 - 3 | 367 | 70 | |
| GenBank mRNA | AK013554.1 | - | 2 - 3 | 319 | 70 | |
| AK028937.1 | - | 2 - 3 | 293 | 70 | ||
| AK028945.1 | - | 2 - 3 | 270 | 70 | ||
| AK030280.1 | - | 2 - 3 | 307 | 70 | ||
| AK036108.1 | - | 2 - 3 | 291 | 70 | ||
| AK134887.1 | - | 2 - 3 | 324 | 70 | ||
| AY029531.1 | - | 2 - 3 | 268 | 70 | ||
| BC003946.1 | - | 2 - 3 | 254 | 3487343 | 70 | |
| CK617511.1 | - | 2 - 3 | 443 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cers4
Genomic Map