Assay Details
| Gene Symbol: Cers4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026058.4 | NP_080334.3 | 6 - 7 | 704 | 73 | |
| GenBank mRNA | AK013554.1 | - | 6 - 7 | 656 | 73 | |
| AK028937.1 | - | 6 - 7 | 630 | 73 | ||
| AK028945.1 | - | 6 - 7 | 607 | 73 | ||
| AK030280.1 | - | 6 - 7 | 644 | 73 | ||
| AK215994.1 | - | 3 - 4 | 157 | 73 | ||
| AY029531.1 | - | 6 - 7 | 605 | 73 | ||
| BC003946.1 | - | 6 - 7 | 591 | 3487343 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cers4
Genomic Map