Assay Details
| Gene Symbol: Taf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081092.1 | NP_001074561.1 | 5 - 6 | 1755 | 89 | |
| GenBank mRNA | AY038601.1 | - | 5 - 6 | 611 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Taf4a
Genomic Map